missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes

Product Code. 10707674
Change view
Click to view available options
Measurement Range:
0.2 to 2%
40 to 800
Unit Size:
5 tests
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10707674 40 to 800 5 tests
10454292 0.2 to 2% Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707674 Supplier Dräger™ Supplier No. 8103381
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Components Measured Acetone
Measurement Range 40 to 800
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C
Tube Type Acetone 40/a
compliance-icons
Product Identifier
  • drager-tubes
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contain: acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.