missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Carbon Monoxide

Product Code. 10262531
Change view
Click to view available options
Measurement Range:
0.3 to 7%
1:2.5 to 150
2 to 50 ppm
5 to 700
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range unitSize
10262531 2 to 50 ppm Pack of 10
12806673 0.3 to 7% Pack of 10
10717284 1:2.5 to 150 Pack of 10
10476871 5 to 700 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10262531 Supplier Dräger™ Supplier No. 6733051
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Carbon Monoxide
Humidity Range 2 to 20mg/L
Measurement Range 2 to 50 ppm
No. of Strokes 10/2
Quantity 10
Response Time 4 min./50 sec.
Temperature Range (English) 32°F to 122°F
Temperature Range (Metric) 0°C to 50°C
Tube Type Carbon monoxide 2/a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.