Learn More
Dräger™ Short-Term Detector Tubes: Chloroform
Detector tubes
Brand: Dräger™ 6728861
Code : NEW
Additional Details : Weight : 0.03000kg Transport : UN number : 3260 Chem class : 8 Pack group : III
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Chloroform | |
2 to 10 | |
10 | |
50°F to 86°F | |
Chloroform 2/a |
9mg/L | |
10 | |
9 min. | |
10°C to 30°C |