missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrogen

Product Code. 10325192
Change view
Click to view available options
Components Measured:
Hydrogen Fluoride 1.5/b (gas or vapor)
Hydrogen Peroxide 0.1/a (gas or vapor)
Measurement Range:
0.1 to 3ppm
15ppm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range unitSize
10325192 Hydrogen Fluoride 1.5/b (gas or vapor) 15ppm Pack of 10
10530663 Hydrogen Peroxide 0.1/a (gas or vapor) 0.1 to 3ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10325192 Supplier Dräger™ Supplier No. CH30301
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Hydrogen Fluoride 1.5/b (gas or vapor)
Measurement Range 15ppm
Quantity 10/Box
compliance-icons
Product Identifier
  • Hydrogen Fluoride 1.5/b
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.