missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Fluoride

Product Code. 12754698
Change view
Click to view available options
Measurement Range:
0.5 to 90ppm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
12754698 0.5 to 90ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12754698 Supplier Dräger™ Supplier No. 8103251
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides a reliable, cost-effective method for short-term, spot measurement of hydrogen fluoride gas concentrations.

The Dräger tube is a glass vial filled with a chemical reagent designed for the detection of hydrogen fluoride. It is used in conjunction with a Dräger gas detection pump, such as the Accuro™ pump, to measure gas concentrations through a colorimetric reaction.

  • Standard Measurement Range: 0.5 to 15 ppm
  • Measurement Time: Approximately 25 seconds
  • Number of Strokes (n): 10
  • Standard Deviation: 20% to 30%
  • Color Change: Blue-violet to yellow
  • Operating Temperature: 10°C to 40°C
  • Operating Humidity: 30% to 80%
  • Reaction Principle: HF + pH-indicator → yellow reaction product
  • Cross-Sensitivity: Mineral acids, such as hydrochloric acid or nitric acid, are also indicated. Basic gases, such as ammonia, can cause negative errors or prevent indication entirely.

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Hydrogen Fluoride 0.5/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 30 to 80%
Measurement Range 0.5 to 90ppm
No. of Strokes 10/2
Quantity 10
Response Time 2 min./25 sec.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.