missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Mercury

Product Code. 12886663
Change view
Click to view available options
Measurement Range:
0.05 to 2mg/m3
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
12886663 0.05 to 2mg/m3 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12886663 Supplier Dräger™ Supplier No. CH23101
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glass reagent tube for the measurement of mercury vapor concentrations using a gas detection pump.

Dräger tube for the detection of mercury vapor. The tube contains a chemical reagent that changes color to indicate concentration.

  • Standard Range: 0.05 to 2 mg/m³
  • Measurement Time: 10 minutes
  • Calibrated Sample Volume: 100 mL
  • Compatible Pumps: Dräger Accuro™ bellows pump, Accuro 2000, or Quantimeter 1000
  • Applications: Determination of concentration peaks, exposure measurements in inhalation areas, leak detection, and analysis of confined spaces such as sewers, shafts, and tanks

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Mercury 0.1/b
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range Less than20mg/L
Measurement Range 0.05 to 2mg/m3
No. of Strokes 40 to 1
Quantity 10
Response Time 10 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.