Learn More
Dräger™ Short-Term Detector Tubes - De Tube Methylene Chloride
Brand: Dräger™ 8103591
Code : 50
Additional Details : CAS Number : 7664-93-9 Weight : 0.09180kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
Methylene Chloride 100/ab | |
20 to 200ppm | |
10 | |
62.6°F to 86°F |
3 to 25mg/L | |
8 | |
7 min. | |
17°C to 30°C |
Safety and Handling
- X10 GAS DETECTOR TUBE DRAEGER 6724601 METHYLENE
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Supplemental information
- MIXTURE LIST-Contains : Amine compounds