missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - n-Octane

Product Code. 10049230
Change view
Click to view available options
Measurement Range:
10 to 300ppm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10049230 10 to 300ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10049230 Supplier Dräger™ Supplier No. 8101691
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Short-term detector tube for the measurement of petroleum hydrocarbons in confined spaces.

The Petroleum Hydrocarbons 10/a detector tube is a glass vial filled with a chemical reagent designed for the determination of air contaminants in the workplace.

  • Detection Principle: Colorimetric chemical sensor (C8H18 + I2O5 → I2); the concentration is directly indicated by the length of the color change from white to brownish-green.
  • Standard Measuring Range: 10 ppm to 300 ppm (for n-octane).
  • Measurement Time: Approximately 1 min.
  • Number of Strokes (n): 2.
  • Sample Volume: 100 mL.
  • Standard Deviation: ±25%.
  • Operating Temperature: 15°C to 30°C.
  • Operating Humidity: 1 to 20 mg H2O/L.
  • Cross-Sensitivity: Indicates various organic and inorganic compounds in addition to n-octane, including n-hexane, n-heptane, iso-octane, n-nonane, perchloroethylene, and CO.
  • Compatibility: Used manually with the Dräger Accuro™ bellows pump or automatically with the Accuro 2000 pump and Quantimeter 1000.
  • Applications: Determination of concentration peaks, exposure measurements in the inhalation area, leak detection, and analysis of air in sewers, shafts, tanks, or other confined spaces.

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Petroleum Hydrocarbons 10/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Measurement Range 10 to 300ppm
No. of Strokes 8
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 41°F to 77°F
Temperature Range (Metric) 5° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.