missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - Phenol

Product Code. 13101166
Change view
Click to view available options
Measurement Range:
1 to 20ppm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
13101166 1 to 20ppm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13101166 Supplier Dräger™ Supplier No. 8101641
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glass reagent tubes designed for the colorimetric measurement of phenol concentrations in air samples using a gas detection pump.

DrägerTubes are glass vials filled with a chemical reagent designed for gas detection. When a 100 mL sample of air is drawn through the tube, the reagent changes color to indicate the presence and concentration of the target substance.

  • Product Type: Short-term tubes
  • Measurement Time: 10 sec to 15 min
  • Configuration: Tubes with one indicating layer
  • Compatibility: Designed for use with Dräger Accuro™ gas detection pump, Accuro 2000, or Quantimeter 1000
  • Applications: Determination of concentration peaks, measurement of exposures in the inhalation area, leak detection, and analysis of air in sewers, shafts, tanks, or other confined spaces

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Phenol 1/b
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 1 to 18mg/L
Measurement Range 1 to 20ppm
No. of Strokes 20
Quantity 10
Response Time 5 min.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.