missing translation for 'onlineSavingsMsg'
Learn More

Draeger Short Term Tubes

Product Code. 10364020
Change view
Click to view available options
Components Measured:
Mercaptan 0.1/a
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
10364020 Mercaptan 0.1/a Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10364020 Supplier Draeger Supplier No. 8103281
  • ONLINE EXCLUSIVE PRICE
    220.46 €
    197.18 € / Pack of 10
    Save 23.28 €
    10% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy to use Draeger tubes are proven to be very cost effective and reliable method for th measurement of gases.

  • Quick and targeted measurements
  • Easy handling either manually using the Draeger-Tube pump accuro or automatically with the Draeger X-act 5000
  • Field of application includes air examinations at the workplace, technical gas analysis in areas of emission/immission concentrations, measurement of compressed air/gases
  • Measures a large number of different gases and vapours

Specifications

Components Measured Mercaptan 0.1/a
Type Short-term Draeger Sampling Tubes
For Use With (Application) Spot Measurement of More That 500 Different Gases
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.