missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dräger™ Detector Tube - Hydrocyanic Acid
Click to view available options
Components Measured:
Hydrocyanic Acid
Unit Size:
Pack of 10
Description
- Sealed glass vial
- Contains solid carrier material and chemical reagent which react to particular gas/vapour
- Provides exact results
- Calibration showed on in form of scale printed on tube
- Applications in industry, fire fighting, diaster prevention, laboratory work and environmental protection
Specifications
Specifications
| Detection Limits | 0.5 to 100ppm |
| Components Measured | Hydrocyanic Acid |
| Material | Glass |
| Quantity | 10/Pk. |
| Tube Type | Detector Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction