missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start PCR Master Mixes

Product Code. 15699374 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Colorless
Green
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
1000 reactions
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color No. of Reactions Unit Size
15699374 Green 1000 Reactions Each
15669374 Colorless 200 Reactions Each
15717536 Colorless 200 Reactions Each
15686154 Colorless 1000 Reactions 1000 reactions
15727536 Colorless 1000 Reactions Each
15679374 Colorless 1000 Reactions Each
15737536 Green 200 Reactions Each
15689374 Green 200 Reactions Each
15606164 Green 1000 Reactions 1000 reactions
15747536 Green 1000 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 15699374 Supplier Thermo Scientific™ Supplier No. K9022
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific DreamTaq Hot Start PCR Master Mix is a ready-to-use 2X reaction mix that includes DreamTaq Hot Start DNA Polymerase, DreamTaq buffer, magnesium, and dNTPs.

Thermo Scientific DreamTaq Hot Start PCR Master Mix, available in colorless and green formats, is a ready-to-use 2X reaction mix that includes DreamTaq Hot Start DNA Polymerase, DreamTaq Buffer, magnesium, and dNTPs.

The master mix allows easy reaction setup and fewer pipetting steps, with only primers, template, and water needing to be added. The DreamTaq Hot Start Green PCR Master Mix includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases.

DreamTaq Hot Start PCR Master Mix is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Simplified workflow and minimized pipetting steps with 2X master mix format
    • Direct loading of PCR products on a gel with green format

Applications:

  • Routine genomic and plasmid DNA amplification
  • Genotyping
  • Colony PCR and clone screening
  • High-throughput PCR screening
  • TA cloning workflows

Recommended Storage

Store at -20C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X DreamTaq Hot Start Green PCR Master Mix, 20 x 1.25 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -5°C to -30°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Quantity 1000 rxns
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Hot Start Built-In Hot Start
No. of Reactions 1000 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Color Green
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.