missing translation for 'onlineSavingsMsg'
Learn More

DreamTaq DreamTaq™ Hot Start PCR Master Mix

Product Code. 15727536 Shop All Thermo Scientific Products
This item is not returnable. View return policy

Product Code. 15727536

Brand: DreamTaq K9012

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free

Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.

Better reaction outcomes: 
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.  
  • Simplified workflow and minimized pipetting steps with 2X master mix format.  
Amplicon size (complex DNA): 6 kb
Fidelity (vs. Taq): 1X

Recommended Storage

Store at -20C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• 20 x 1.25 mL DreamTaq Hot Start PCR Master Mix (2X)
• 20 x 1.25 mL Nuclease-free water

Store at -5 to -30°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Shipping Condition Wet or Dry Ice
Hot Start Built-In Hot Start
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.