missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq PCR Master Mixes (2X)

Product Code. 11816843 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Colorless
Green
Quantity:
4 x 1.25 mL
20 x 1.25 mL
Unit Size:
1000 reactions
200 reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
11816843 Green 20 x 1.25 mL 1000 reactions
11896833 Colorless 4 x 1.25 mL 200 reactions
15787516 Colorless 4 x 1.25 mL 200 reactions
11806843 Colorless 20 x 1.25 mL 1000 reactions
15797516 Colorless 20 x 1.25 mL 1000 reactions
11813933 Green 4 x 1.25 mL 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11816843 Supplier Thermo Scientific™ Supplier No. K1082
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.

Thermo Scientific DreamTaq PCR Master Mix (2X), available in colorless and green formats, is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR setup.

The master mix retains all features of DreamTaq DNA Polymerase and is capable of robust amplification up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.

DreamTaq Green PCR Master Mix (2X) additionally includes 2X DreamTaq Green Buffer with a density reagent and two dyes for monitoring electrophoresis progress.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications:

  • Routine genomic and plasmid DNA amplification
  • Genotyping
  • Colony PCR and clone screening
  • High-throughput PCR screening
  • TA cloning workflows
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X DreamTaq Green PCR Master Mix, 20 x 1.25 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -20°C.
GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Quantity 20 x 1.25 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start No
No. of Reactions 1000 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Color Green
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.