missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, indicating (CaSO{4})

Product Code. 15491857
Change view
Click to view available options
Quantity:
2270 g
Unit Size:
2270g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491857 2270 g 2270g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491857 Supplier Thermo Scientific Alfa Aesar Supplier No. 043063.=A
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

Drierite™, indicating (CaSO{4}) is used as a desiccant. It is used as a drying agent utilized for the efficient and rapid drying of air, industrial gases, refrigerants, organic liquids and solids. Further, it is used to maintain a dry atmosphere in storage spaces, vaults, commercial packages, and other enclosures. In addition to this, it is involved in the protection of hygroscopic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Granular
Quantity 2270 g
Odor Odorless
Solubility Information Soluble aq. soln..
Sensitivity Hygroscopic
Grade Laboratory
Chemical Name or Material Drierite
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Drierite, indicating
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Calcium sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.