missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, With indicator, 8 mesh, Honeywell™ Fluka™

Product Code. 15662400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
454 g
2.3 kg
12 x 454 g
Packaging:
Fiber Drum
Glass Bottle
Unit Size:
2.3kg
454g
Pack of 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15662400 454 g Glass Bottle 454g
15652400 2.3 kg Glass Bottle 2.3kg
15642400 12 x 454 g Fiber Drum Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15662400 Supplier Honeywell-Fluka Supplier No. 238988454G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With indicator, 8 mesh, Drierite™ is a trademark of W. A. Hammond Drierite Company, Ltd.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Material Calcium Sulfate
Quantity 454 g
Linear Formula CaSO4
Mesh Size 8mesh
UN Number UN3077
Packaging Glass Bottle
Formula Weight 136.14g/mol
Chemical Name or Material Calcium Sulfate, With indicator
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.