missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, without indicator, 20-40 mesh

Product Code. 10582041 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
454 g
2.3 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
2.3kg
454g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582041 454 g Glass Bottle 454g
10125230 2.3 kg Plastic Bottle 2.3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10582041 Supplier Thermo Scientific Supplier No. 219074540
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifica

Melting Point 1450.0°C
Physical Form Granular
Quantity 454 g
Fieser 01,107
Merck Index 15, 1708
Packaging Glass Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol. SiO2 and insolubles: 0.15%
R2O 0.14%
Formula Weight 136.14
Chemical Name or Material Drierite™, 20-40 mesh

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.