missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, without indicator, 4 mesh

Product Code. 10711881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711881 2 kg Plastic Bottle 2kg
10721881 500 g Glass Bottle 500g
11934251 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711881 Supplier Thermo Scientific Acros Supplier No. 219030020
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Physical Form Granular
Quantity 2 kg
Packaging Plastic Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
IUPAC Name calcium sulfate
Formula Weight 136.14
Chemical Name or Material Drierite™

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.