missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Drop-boy Dropping Bottles

Product Code. 10445951 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10445951 25 mL Each
10438831 10 mL Each
10056901 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10445951 Supplier Buerkle™ Supplier No. 03070025
  • ONLINE EXCLUSIVE PRICE
    1.50 €
    1.31 € / Pack of 1
    Save 0.19 €
    12% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Buerkle™ Drop-boy Dropping Bottles for accurate, reliable and repeatable dispensing of reagents.

  • HDPE
  • Drop dispenser and separate screw cap
  • Closure included
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 25 mL
Color White
Diameter (English) 1.22 in.
Diameter (Metric) Inner Neck 13 mm
Includes Drop dispenser and separate screw cap
Height (Metric) With Closure 103 mm
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 65 mm
Height (English) 2.56 in.
Diameter (Metric) 31 mm
Closure Type Drip Closure
Autoclavable Not Autoclavable
Capacity (English) 0.85 oz.
Shape Round
Disposable Reusable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.