missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Drop-boy Dropping Bottles

Product Code. p-8001977 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10438831 10 mL Each
10445951 25 mL Each
10056901 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10438831 Supplier Buerkle™ Supplier No. 03070010
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Buerkle™ Drop-boy Dropping Bottles for accurate, reliable and repeatable dispensing of reagents.

  • HDPE
  • Drop dispenser and separate screw cap
  • Closure included
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 10 mL
Includes Drop dispenser and separate screw cap
Material HDPE
Closure Included Yes
Height (Metric) 50 mm
Diameter (Metric) 24 mm
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.