Learn More
Thermo Scientific™ μDrop™ and μDrop Duo Plates
Product Code. 16638987 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
μDrop Duo Plate
μDrop Plate
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16638987 μDrop Duo Plate Each
11899143 μDrop Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16638987 Supplier Thermo Scientific™ Supplier No. N12391M2
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ μDrop and μDrop Duo plates are used for low volume (2-10 μl) photometric DNA, RNA and protein quantitation and purity analysis. The μDrop plates are compatible with Thermo Scientific Varioskan LUX and Multiskan SkyHigh microplate readers. The ready-made protocols in SkanIt software or Multiskan SkyHigh user interface make quantification fast and easy.
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Up to 16 (μDrop Plate) or 32 (μDrop Duo Plate) sample positions for quick and easy measurement of sample volumes down to 2 μl
  • Compatible with an eight-channel pipette for sample addition
  • Easy to clean after measurement
  • Measurement slot for a rectangular cuvette
  • Ready-made Multiskan SkyHigh user interface and SkanIt Software protocols for nucleic acid and protein analysis
  • Compatible with Multiskan Sky, Multiskan SkyHigh and Varioskan LUX microplate readers
TRUSTED_SUSTAINABILITY
Specifications
Product Type μDrop Duo Plate
For Use With (Equipment) Varioskan LUX, Multiskan Sky, Multiskan SkyHigh
Plate Type μDrop Duo Plate
Capacity (Metric) 2 to 10μL (low-volume function)
For Use With (Application) Nucleic acid and protein concentration measurement
No. of Positions 32