missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Dropping Funnel with Pressure Equalization

Product Code. 10573814
Change view
Click to view available options
Capacity (Metric):
100 mL
Stopcock:
ST 14/23
ST-PTFE 29/32
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Stopcock Unit Size
10573814 100 mL ST 14/23 -
10688115 100 mL ST-PTFE 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10573814 Supplier Witeg™ Supplier No. 1 850 100
  • ONLINE EXCLUSIVE PRICE
    82.00 €
    70.23 € / Pack of 1
    Save 11.77 €
    14% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this funnel to slowly add air-sensitive reagents in a sealed environment. Witeg™ Dropping Funnel with Pressure Equalization facilitates the drop-wise addition of reagents.

  • Pressure equalizing.
  • Graduated.
  • Pack: 2 pieces.

Specifications

Capacity (Metric) 100 mL
Material Glass
Description Pressure equalizing
Graduated Yes
Graduations 2mL
Stopcock ST 14/23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.