Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Product Code. 10550152 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
2.5cm narrow plastic shell vials
2.8cm wide plastic shell vials
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10550152 2.8cm wide plastic shell vials Pack of 1000
10346221 2.5cm narrow plastic shell vials Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10550152
Spotlight Offers
Use promo code 28038
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
For Use With (Application) Minimizes evaporation, keeps food fresh longer
For Use With (Equipment) 2.8cm wide plastic shell vials
Includes Superior barrier
Quantity 1000 Case
Material Cellulose Acetate
Type BuzzPlugs Bottle Closure