missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Drug of Abuse Rapid Test - Alcohol Test Card

Product Code. 31454318 Shop All MP Biomedicals Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form
31454318 Ready-to-Use (RTU)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31454318 Supplier MP Biomedicals Supplier No. 07RD4014
  • ONLINE EXCLUSIVE PRICE
    3.23 €
    3.08 € / 1 tests
    Save 0.15 €
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alcohol Test Card is an immunochromatography based one step in vitro test.

The Drug of Abuse Rapid Test - Alcohol Test Card is based on the high specificity of alcohol oxidase (AlOx) for ethyl alcohol in the presence of peroxidase and enzyme substrate such as tetramethylbenzidine (TMB). The distinct color on reactive pad could be observed in less than 20 seconds after the urine samples migrates over the reaction pad with the ethyl alcohol concentration greater than 0.04%. It should be pointed out that other alcohols such as methyl, propanyl and allyl alcohol would develop the similar color on the reactive pad. However, these alcohols are not normally present in urine.

  • CE-marked
  • It is designed for qualitative determination of Alcohol in human urine specimens.
  • For this reference, the parameter and the cut-off value are: Alcohol 40 mg/dL.

Specifications

CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes Alcohol
Format Test Card
Form Ready-to-Use (RTU)
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.