missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Drug of Abuse Rapid Test - Cup 7 parameters (BAR300/BUP10/BZD300/COC300/MTD300/OPI300/THC50)

Product Code. 31454340 Shop All MP Biomedicals Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form
31454340 Ready-to-Use (RTU)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31454340 Supplier MP Biomedicals Supplier No. 07RD720706
  • ONLINE EXCLUSIVE PRICE
    6.30 €
    6.00 € / 1 tests
    Save 0.30 €
    4% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Drug of Abuse Rapid Test - Cup 7 parameters is an immunochromatography based one step in vitro test.

The Drug of Abuse Rapid Test - Cup is based on the principle of specific immunochemical reaction between antibodies and antigen to analyze particular compounds in human urine specimen. The assay relies on the competition for the binding antibody. When drug is present in the urine specimen, it competes with drug conjugate for the limited amount of antibody-dye conjugate. When the amount of drug is equal or more than the cut-off, it will prevent the binding of drug conjugate to the antibody. Therefore, a positive urine specimen will not show a colored band on the test line zone, indicating a positive result, while the presence of a colored band indicates a negative result. A control line is present in the test window to work as procedural control. This colored band should always appear on the control line zone if the test device is stored in good condition and the test is performed appropriately.

  • CE-marked
  • Drug detection in human urine sample
  • Tailor-made tests with customized combinations
  • More than 30 parameters: amphetamine, benzodiazepine, barbiturate, buprenorphine, cocaine, EDDP, ketamine, methamphetamine, MDMA, methadone, opiates, oxycodone, pencyclidine, THC, propoxyphene, tramadol, tricyclic antidepressant, methylphenidate, fentanyl, synthetic cannabis, clonazepam, cotinine, alcohol Adulteration parameters: oxidants, bleach, pyridinium chlorochromate, nitrite, creatinine, pH and specific gravity
  • For this reference, the parameters and cut-off values are: Barbiturate 300 ng/mL, Buprenorphine 10 ng/mL, Benzodiazepine 300 ng/mL, Cocaine 300 ng/mL, Methadone 300 ng/mL, Opiate 300 ng/mL, THC 50 ng/mL.

Specifications

CE Marker Self-declared
Clia Complexity Not Applicable
Detectable Analytes Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methadone (MTD), Opiates (OPI), Cannabinoids (THC)
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.