Learn More
Manutan Drum key, steel
Brand: Manutan A350888
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Easily plugs or removes bungs up to 2 in diameter from steel or plastic drums without causing any damage. Compact and lightweight, can be carried on the person or in a toolbox. Fitted with an extractor pin for effortlessly peeling off metal sealing caps.Steel