missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Dry-Block Heater/Cooler

Product Code. 17795019
Change view
Click to view available options
Tube Size:
1.5 mL
2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Size unitSize
17795019 1.5 mL Each
17705029 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17795019 Supplier Grant Instruments™ Supplier No. PCH2 12V
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Grant Instruments PCH-1/PCH-2/PCH-3 personal benchtop cooler/heaters are ideal for laboratory applications requiring cooling and heating in microtubes within the temperature setting range of – 10°C to +100°C.

The PCH cooler/heaters enable a stable temperature to be maintained during room temperature fluctuations.
  • The ingenious block construction, combined with a powerful Peltier module, produces very rapid cooling and heating for swift change of application and increased flexibility. In addition, the Peltier effect produces efficient cooling and heating for small thermal loads without the use of a refrigeration system.
  • The dry temperature control system maintains a clean working environment, reducing the chance of contamination or sample dilution. A Peltier system, without the vibration of refrigeration or the turbulence of a waterbath, is also ideal for all incubations where stable conditions are critical.
  • The PCH is a very effective tool for DNA sample preparation for denaturing electrophoresis. The exceptionally rapid heat up enables the denaturation temperature of 95°C to be reached swiftly, the timer will signify denaturation time-up, and the samples can be rapidly cooled again ready to load the gels.
  • Cooling/heating setting range from – 10ºC to 100°C, with very rapid cool down and heat-up times
  • Stability ± 0.1°C, uniformity ±0.25°C at 37°C
  • Choice of two models; capacity for up to 32 microtubes in a combination of two sizes: 12 x 1.5 mL plus 20 x 0.5 mL (PCH-1) or up to 20 microtubes of one size: 20 x 1.5 mL tubes (PCH-2), up to a total of 20 x 2 mL microtubes (PCH-3)
  • Convenient integral reaction timer with audible alarm
  • Dry temperature control system maintains a clean and vibration-free working environment
  • A 2-line LCD display clearly indicates both set and actual values for temperature and time. Works in combination with simple push buttons for easy set-up.
  • Flexible tube sizes and rapid heat-up/cool-down times enables swift change of application

Applications:

Life science – storing frozen restriction enzymes, nick translations, ligation reactions, restriction digests, protein solubilisation for PAGE, warm incubation of microcentrifuge tubes for hybridisation, enzyme reactions and deactivations.

Clinical – cooling blood samples prior to coagulation testing.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Dry-block heater/cooler for 20 x 1.5 mL microtubes -10°C to +100°C (ambient -30°C minimum)
Holds 20 Microtubes
Plug Type US, EU, UK, AUS Plug
Temperature Range (Metric) -10°C to +100°C (Cooling/Heating Setting Range)
Type Dry-Block Heater/Cooler
Display 2 line x 16 character LCD
Model PCH-2 12 V
No. of Blocks 1
Temperature Stability Stability ±0.1°C, uniformity ±0.25°C at 37°C
Timer 1 min. to 96 hr., Non-stop
Tube Size 1.5 mL
Warranty 2 years
Dimensions (L x W x H) 32 x 30 x 25 cm
Shipping Weight (Metric) 4 kg
Weight (Metric) 3.2 kg
Voltage 12 V
Wattage 55 W
Amperage 4.4 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.