missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dry-Ready™ RT-qPCR Kit

Product Code. 31722750
Change view
Click to view available options
Quantity:
200 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31722750 200 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31722750 Supplier Invitrogen™ Supplier No. A40006906
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Experience the simplicity and efficiency of the air drying process by gently heating materials at moderate temperatures in a simple ventilated oven.

The Invitrogen Dry-Ready RT-qPCR Kit is an innovative solution designed for preparation of hot air-dried RT-qPCR assays. A simple and fast air-drying method surpasses traditional lyophilization in both speed and simplicity, while also enabling assay storage and shipment at room temperature.

The kit includes Dry-Ready Platinum II DNA Polymerase, Dry-Ready SuperScript III Flash Reverse Transcriptase, Dry-Ready RNaseOUT Recombinant Ribonuclease Inhibitor, Dry-Ready dNTP Mix, 10X Dry-Ready Reaction Buffer with exipients, and MgCl2 for a suite of essential components for one-step RT-qPCR applications.

The Dry-Ready RT-qPCR Kit represents a significant advancement in one-step RT-qPCR technology, offering a solution that combines innovation with reliability. Features include:

  • Complete set of components for RT-qPCR assay drying and downstream applications
  • Dry-Ready SuperScript III Flash Reverse Transcriptase enables reverse transcription in just one minute
  • Dry-Ready Platinum II DNA Polymerase helps ensure high reliability and increased tolerance to inhibitors
  • Enhanced stability, specifically optimized for air drying applications

Air-drying technology

Utilizing this kit is straightforward and efficient: users need only mix the RT-qPCR assay components and aliquot into single reaction volumes. The mixture is then dried in an oven. This uncomplicated process results in a stable assay that can be stored at room temperature for at least 12 months without loss of efficiency.

By incorporating all these features, the Dry-Ready RT-qPCR Kit stands as an innovative solution in the field of molecular diagnostics and research. Its unique combination of enzyme stability, ease of use, and long-term storage capability at room temperature positions it as a valuable asset for laboratories looking to advance their RT-qPCR capabilities while streamlining their processes.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

•Dry-Ready™ Platinum™ II DNA Polymerase, 24 μL
•Dry-Ready™ SuperScript™ III Flash Reverse transcriptase, 20 μL
•Dry-Ready™ RNaseOUT™, 40 μL
•Dry-Ready™ dNTP Mix, 80 μL
•MgCl2, 40 μL
•Store at -20°C.

Detection Method qPCR, RT-PCR
Form Liquid
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Polymerase Platinum™ II Taq Hot-Start DNA Polymerase
Reaction Speed Fast
Label or Dye Unlabeled
No. of Tests 200 Reactions
Product Type RT-qPCR Kit
Quantity 200 Reactions
Reverse Transcriptase SuperScript™ III Flash Reverse Transcriptase
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Sample Type RNA & DNA
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.