Learn More
Heinz Herenz Dry Swabs
Product Code. 15283728
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 150
Product Code. Sterility Unit Size
15283728 Sterile Pack of 150
1 options
Product Code. 15283728 Supplier Heinz Herenz Supplier No. 1020005
  • ONLINE EXCLUSIVE PRICE
    75.50 €
    50.51 € / Pack of 150
    Save 24.99 €
    33% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dry swabs with medium/ transport swabs. Sterile with Amies medium. Plastic applicator with viscose/rayon head, sterilized by radiation.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Product Type Applicator Swab and Tube, Swab
Format Swab
Sterility Sterile
Applicator Material Polystyrene
Tip Material Polystyrene
Tip Type Viscose
Media Type Liquid Amies, Amies Medium
Color White
No. per Pack 150Pack
Quantity 150Pack
Material Plastic