Learn More
Lenz Laborglasintrumente Drying Tubes, Bent, with NS Cone
Product Code. 10030241
Change view
Click to view available options
Standard Taper Joint No.:
NS 24/29
Unit Size:
Each
Product Code. Standard Taper Joint No. Unit Size
10030241 NS 24/29 Each
1 options
Product Code. 10030241 Supplier Lenz Laborglasintrumente Supplier No. 05337003
  • ONLINE EXCLUSIVE PRICE
    35.00 €
    22.67 € / Pack of 1
    Save 12.33 €
    35% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Drying tubes with NS cone
  • Bent (75°)
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Standard Taper Joint No. NS 24/29