missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-32 Matrix Standard Kit (Dye Set F)

Product Code. 15899796 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
8 Runs
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15899796 8 Runs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15899796 Supplier Applied Biosystems™ Supplier No. 4345831
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Contains one set of matrix standard DNA fragments with unique fluorescent dye labels, enough for 8 runs

The matrix standard set DS-32 is used to generate the 'multicomponent matrix' required when analyzing 5-FAM™, JOE™, NED™, and ROX™ labeled DNA fragments on the Applied Biosystems™ 31xx, 3500 and 3730 Series Systems.

The matrix standard set DS-32 is used to generate the 'multicomponent matrix' required when analyzing 5-FAM™, JOE™, NED™, and ROX™ labeled DNA fragments on the Applied Biosystems™ 3500 and 3730 Series Systems. The Data Collection Software uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

Supports 4-Dye Human Identity applications (i.e., AmpFLSTR™ kits), StockMarks™, and Plant/Microbial AFLP assays.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains one set of matrix standard DNA fragments with unique fluorescent dye labels. Enough for 8 runs. Store the kit at 2 - 8°C. Do not freeze.
Format Kit
For Use With (Equipment) SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3130 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye FAM, JOE, NED, ROX
Product Type DS-32 Matrix Standard Kit
Quantity 8 Runs
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.