missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ DSPC

Product Code. 18706295
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18706295 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18706295 Supplier R&D Systems™ Supplier No. 7943/100
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Neutral phospholipid; component of lipid nanoparticles (LNPs)

DSPC is a phosphatidylcholine, a type of neutral phospholipid with two saturated long-chains (18:0). Commonly used in the generation of micelles, liposomes, lipid nanoparticles (LNPs) and other types of artificial membranes. DSPC is used in formulation of LNPs for nonviral drug delivery systems.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphocholine
CAS 816-94-4
Quantity 100 mg
Product Line Tocris
Target Lipid Nanoparticles
Molecular Formula C44H88NO8P
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.