missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DSS (disuccinimidyl suberate), No-Weigh™ Format

Product Code. 15971252 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
8 x 2 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15971252 10 x 2 mg Pack of 10
11831255 8 x 2 mg Pack of 8
10138353 50 mg 50mg
10540434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15971252 Supplier Thermo Scientific™ Supplier No. A39267
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Crosslink amines to amines with disuccinimidyl suberate composed of NHS-ester groups with a membrane-permeable, mid-length spacer for intracellular protein crosslinking.

Thermo Scientific™ Pierce DSS, also called disuccinimidyl suberate, is noncleavable and membrane permeable crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm.

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Amine reactive sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
  • Membrane-permeable, allowing for intracellular crosslinking
  • High purity, crystalline reagent can be used to create high-purity conjugates
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO); compare to BS3 (Sulfo-DSS)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DSS
Molecular Weight (g/mol) 368.35
PEGylated No
Spacer Arm Length 11.4 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 10 x 2 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less
What is the protocol for crosslinking IgG to Dynabeads from the Dynabeads Protein G Immunoprecipitation Kit (Cat. No. 10007D) using DSS (disuccinimidyl suberate)?

For crosslinking the antibody to Protein G Dynabeads, please refer to the crosslinking protocol with BS3 (bis(sulfosuccinimidyl)suberate) or Sulpha-DSS, which is an analogue of DSS. The only difference between these two molecules is that DSS is water insoluble and needs to be solubilized in DMSO before use. You can find the protocol at the following link.

Can you provide the shelf-life for DSS (disuccinimidyl suberate)?

DSS (disuccinimidyl suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

How does a product with "No-Weigh" format reduce my concern over reagent stability?

The No-Weigh product format, where small amounts of reagent are pre-weighed and packaged separately, prevents the contamination and loss of reagent reactivity over time associated with repetitive opening and closing of a single vial. This format enables use of a fresh vial of reagent each time and eliminates the hassle of weighing out small amounts of reagent. Please view additional information and the video at: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-labeling-crosslinking/no-weigh-packaging.html

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.