missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DTNB

Product Code. 16489447
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489447 5 g 5g
16481569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489447 Supplier MedChemExpress Supplier No. HY15915
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DTNB (Ellman’s Reagent) is a chemical used to quantify the number or concentration of thiol groups.

Specifications

Quantity 5 g
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Solubility DMSO : ≥ 150 mg/mL (378.45 mM)
CAS 69-78-3
Molecular Weight (g/mol) 396.35 g/mol
Synonym Ellman’s Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.