missing translation for 'onlineSavingsMsg'
Learn More

Melford DTT

Product Code. 30018219
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
1g
25g
50g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018219 5 g 5g
30018218 25 g 25g
30018216 10 g 10g
30018217 100 g 100g
30018220 50 g 50g
30018215 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018219 Supplier Melford Supplier No. D110005.0
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Protective reagent for maintaining -SH groups in reduced state.

Protective reagent for maintaining -SH groups in reduced state. DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.

Specifications

CAS 12-3-3483
Molecular Formula C4H10O2S2
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.