missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dU Phosphoramidite, standard grade, serum vial bottle

Product Code. 17977132 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μM
1 g
Unit Size:
100µmol
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17977132 1 g 1g
18044090 100 μM 100µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17977132 Supplier Thermo Scientific™ Supplier No. 27173802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis.

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
Full chemical name: 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Molecular formula: C39H47N4O8P
Base: dU
Formula weight: 730.8 g/mol
CAS number: 109389-30-2
HPLC Purity: ≥98.0%; conforms to reference
31P NMR Purity: ≥98.0%; conforms to reference

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type dU CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Deoxyuridine (dU)
Shipping Condition Room Temperature
Purity 98%
Quantity 1 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type DNA
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.