Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Produktkod. 11452107
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Product Type:
D1196, 96-Well
D12384, 384 WELL PCR PLATE
Förpackningsstorlek
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Product Type unitSize
11452107 D12384, 384 WELL PCR PLATE Each
11401997 D1196, 96-Well Each
11411997 D1196, 96-Well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11452107 Supplier Fisherbrand™ Supplier No. D12384
Only null left
Add to basket
Add to basket
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial. Fisherbrand™ Anodized Dual Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
  • Excellent heat transfer
  • Anodized to resist thermal shock
  • For holding test tubes for digital heaters
  • Available to fit a range of tube counts and sizes
Specifications
For Use With (Equipment) PCR
Product Type D12384, 384 WELL PCR PLATE
Quantity 1 CT
Weight 1 kg