Learn More
Jenway™ Dual Cell Holder
Product Code. 11474939
Change view
Click to view available options
Type:
Dual Cell Holder
Unit Size:
Each
Product Code. Type unitSize
11474939 Dual Cell Holder Each
1 options
Product Code. 11474939 Supplier Jenway™ Supplier No. 8307045
Accommodates 10mm square cuvettes and 12.7mm diameter tubes. Jenway™ Dual Cell Holder is exclusivley for use with 6320D Spectrophotometer
  • For use exclusively with Model 6320D Visible Spectrophotometer
  • Holds 10mm square cuvettes and 12.7mm diameter tubes
Specifications
Type Dual Cell Holder
For Use With (Equipment) Jenway 6320D Visible Spectrophotometer
Holds 2 x 10 mm cuvettes and 12.7 mm dia tubes