Learn More
Thermo Scientific™ DualSpin Rotor Body
Product Code. 15580131 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
accuSpin™ 8C Benchtop Centrifuge
Unit Size:
1 unit
Product Code. For Use With (Equipment) Unit Size
15580131 accuSpin™ 8C Benchtop Centrifuge 1 unit
1 options
Product Code. 15580131 Supplier Thermo Scientific™ Supplier No. 75008813
  • ONLINE EXCLUSIVE PRICE
    655.00 €
    566.10 € / Pack of 1
    Save 88.90 €
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Run common clinical IVD blood tubes, gel tubes, 10 mL standard syringes and 15 mL conical tubes with the Thermo Scientific™ DualSpin™ rotor.
TRUSTED_SUSTAINABILITY
Specifications
Material Lightweight Composite
Type Rotor Body
For Use With (Equipment) accuSpin™ 8C Benchtop Centrifuge