missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dulcitol, 97%

Product Code. 11418337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418337 25 g 25g
11428337 100 g 100g
11438337 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418337 Supplier Thermo Scientific Chemicals Supplier No. A18402.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 608-66-2 | C6H14O6 | 182.172 g/mol

Dulcitol is important in studies related to cataract formation, hepatosplenμly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dulcitol is important in studies related to cataract formation, hepatosplenµly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

Solubility
Soluble in water (30g/L at 14°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00064288
InChI Key FBPFZTCFMRRESA-GUCUJZIJSA-N
Synonym dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
PubChem CID 11850
ChEBI CHEBI:16813
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 187°C to 191°C
Density 1.47
Boiling Point 275°C to 280°C (1 mmHg)
Quantity 25 g
Beilstein 1721903
Sensitivity Hygroscopic
Merck Index 14,4332
Solubility Information Soluble in water (30g/L at 14°C).
Formula Weight 182.17
Percent Purity 97%
Chemical Name or Material Dulcitol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.