missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dulcitol, 99+%

Product Code. 10608254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
10608254 500 g Glass bottle 500g
10279213 25 g Glass bottle 25g
10569430 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10608254

Brand: Thermo Scientific Chemicals 117705000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-66-2 | C6H14O6 | 182.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064288
InChI Key FBPFZTCFMRRESA-GUCUJZIJSA-N
Synonym dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
PubChem CID 11850
ChEBI CHEBI:16813
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 188°C to 189°C
Color White
Boiling Point 275°C to 280°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 g
Beilstein 01, 544
Merck Index 15, 4362
Solubility Information (3% in water) Colorless
Formula Weight 182.17
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Dulcitol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.