missing translation for 'onlineSavingsMsg'
Learn More

Dulcitol, TRC

Product Code. 30007875
Change view
Click to view available options
Quantity:
100 mg
1 g
100 g
Unit Size:
100g
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007875 100 g 100g
30004075 1 g 1g
30008529 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007875 Supplier Toronto Research Chemicals Supplier No. TRCD720500100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6 H14 O6
Molecular Weight (g/mol) 182.17
Synonym Dulcite, Dulcitol, Dulcose, Euonymit, Melampyrin, Melampyrit, NSC 1944
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)CO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5+,6-
Formula Weight 182.079
Chemical Name or Material Dulcitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.