missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Dunlop™ Hygrade Boots

Product Code. 15743109
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15743109 43 Each
12004475 41 Each
12084465 39 Each
12074465 38 Each
12044475 45 Each
12034475 44 Each
12094465 40 Each
12014475 42 Each
12054475 46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15743109 Supplier Euro Protection™ Supplier No. 9HYGR43
  • ONLINE EXCLUSIVE PRICE
    60.92 €
    23.69 € / Pack of 1
    Save 37.23 €
    61% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Euro Protection™ Hygrade 9HYGR Safety Boots are constructed of PVC and nitrile and are suitable for use in the food industry.

  • Suitable for food industry.
  • PVC upper.
  • Nylon lining.
  • PVC / Nitrile outsole.

Specifications

Certifications/Compliance CE Certified, EN ISO 20347
Color White, Green
Footbed Material PVC
For Use With (Application) For industrial use, food companies and agricultural work
Liner Material Nylon
Material PVC
Size 43
Sole Features Green color, anti-slip sole
Sole Material PVC
Style Non-safety
Resistant To Anti-slip and waterproof
Waterproof Yes
Product Type Controlled Environments Boots
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.