Learn More
Vileda Professional™ DuoPress™ Wringer
Product Code. 17852486
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
17852486 1 Ea. Each
1 options
Product Code. 17852486 Supplier Vileda Professional™ Supplier No. 168207
  • ONLINE EXCLUSIVE PRICE
    290.00 €
    250.87 € / Pack of 1
    Save 39.13 €
    13% Off
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The CE Trolley range has been specifically designed for use in controlled environments where maximum contamination control is needed. This DuoPress™ wringer is part of the CE Bucket Trolley line for wringing of flat mops.
  • For use on the CE Bucket Trolley with the bucket method
  • 100% polypropylene chassis and stainless steel lever
  • Effectively wrings CE Duo™ mops
  • Ergonomic lever for maximum pressure with minimal effort
  • Base of press support dropped 10 mm to create additional space for mops, making it easier to wring
Specifications
Product Type Mop Wringer
Handle Material Polypropylene and Stainless steel
Includes Wringer base, Support plate and Wringer
Quantity 1 Ea.