missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ DuoSet ELISA Ancillary Reagent Kit 2

Product Code. p-200085175
Change view
Click to view available options
Quantity:
1 Kit (5 x 384-well Plates)
1 Kit (for 5 Plates)
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762154 1 Kit (for 5 Plates) 1 set
30041814 1 Kit (5 x 384-well Plates) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18762154 Supplier R&D Systems™ Supplier No. DY008B
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.