missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ DuoSet ELISA Ancillary Reagent Kit 3
Click to view available options
Quantity:
1 Kit (5 x 384-well Plates)
1 Kit (for 5 Plates)
Unit Size:
1 set
Pack of 5
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction