missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Duotest™ pH Paper Refill

Product Code. 10004700 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
1 to 12
3.5 to 6.8
5 to 8
7 to 10
Unit Size:
Pack of 3
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
10004700 3.5 to 6.8 Pack of 3
10499860 7 to 10 Pack of 3
10799671 1 to 12 Pack of 3
10666561 5 to 8 Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10004700 Supplier Macherey-Nagel™ Supplier No. 90313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Refills give the user the best possible accuracy in pH measurement.

  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colors for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values

Specifications

pH Range 3.5 to 6.8
Format Refill
Quantity 3 Pack
Type pH Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.