missing translation for 'onlineSavingsMsg'
Learn More

Manutan Duplex sling (flat choker), 1000kg capacity, violet, 1m length

Product Code. 12878739
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12878739 Supplier Manutan Supplier No. A340275

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for lifting pipes and bundles, can also be used as basket sling. Two ply webbing and steel 'D' link end fittings, these will weave through each other to form a tight 'choke' around load.

  • Violet
  • Vertical lift SWL: 1000kg
  • Width: 50mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.