missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ Buffer pH 1.68, Hamilton™
Click to view available options
Quantity:
500 mL
Description
- Buffer solutions provide pH stability of up to 5 years
Specifications
pH | 1.68 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
Safety and Handling
Product Identifier
- pH Buffer 1.68
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'