missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper

Product Code. 10179350 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
500 mL
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10179350 2000 mL Each
15231999 1000 mL Pack of 10
15221999 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10179350 Supplier DWK Life Sciences Supplier No. 247016302

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aspirator bottle with ground joint glass side-arm.

Aspirator bottles, ground glass side-arm fro dispensing of liquids. Complying with ISO 4796, these aspirators have a ground socket side-arm and plain neck to accept rubber stoppers. Compatible stopcocks assemblies are available separately.

  • Outlet facilitates attachment of ground jointed stopcock
  • With plain unground neck for rubber stopper and bottom ground joint outlet
  • Made of virtually inert, chemically resistant DURAN™ glass

Specifications

Capacity (Metric) 2000 mL
For Use With (Application) Storage and dispensing of liquids and reagents
Material Borosilicate Glass (3.3)
No. per Pack 1/Pk.
Neck Finish Standard Taper Joint Bottom 19/26
Diameter (Metric) 134 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.